missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iron sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Brand:  Alfa Aesar™ 040856.KS

Code : NEW

Additional Details : CAS Number : 7439-89-6 Weight : 0.01000kg

Product Code. 15434795

  • £434.07 / Each


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Specifications

Iron sputtering target
76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
99.95%
MFCD00010999
1 each
7439-89-6
(metals basis)
Videos
SDS