missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iron slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.95% (metals basis)

Brand:  Alfa Aesar™ 042383.22

Code : NEW

Additional Details : CAS Number : 7439-89-6 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11364508

  • £112.30 / 100g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Iron slug
7439-89-6
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
MFCD00010999

Specifications

Iron slug
100g
7439-89-6
99.95%
MFCD00010999
≈1.58g/slug
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
Slug
(metals basis)
Videos
SDS