missing translation for 'onlineSavingsMsg'
Learn More

Iron powder, 99.5%, Alfa Aesar™

Apparent density 2.0-3.0g/cm3

Brand:  Alfa Aesar 010214.36

Code : NEW

Additional Details : CAS Number : 7439-89-6 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15465925

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Iron
7439-89-6
99.5% (Metals basis)
MFCD00010999
Approximately 98% spherical, 2% fragments
Spherical Powder
Fe

Specifications

Iron
Approximately 98% spherical, 2% fragments
Gray powder
99.5% (Metals basis)
MFCD00010999
500g
7439-89-6
Spherical Powder
Fe
Gray
Videos
SDS