missing translation for 'onlineSavingsMsg'
Learn More

Iron powder, 98% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 039813.36

Code : NEW

Additional Details : CAS Number : 7439-89-6 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15431577

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Iron
7439-89-6
98% (Metals basis)
MFCD00010999
Reduced
Powder
Fe

Specifications

Iron
Reduced
Gray powder
98% (Metals basis)
MFCD00010999
500g
7439-89-6
Powder
Fe
Gray
Videos