missing translation for 'onlineSavingsMsg'
Learn More

Iron powder, 98% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4408732
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
7439-89-6
Fe
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
This item is not returnable. View return policy
7439-89-6
Fe
55.85
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray powder
Density 7.87g/cm3 at 20°C
Assay Percent Range 98% (Metals basis)
MDL Number MFCD00010999
Quantity 250 g
Mesh Size -325 Mesh
Form Powder
Chemical Name or Material Iron, Reduced

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'