missing translation for 'onlineSavingsMsg'
Learn More

Iron powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407463
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
This item is not returnable. View return policy
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99% (metals basis)
Quantity 250 g
Mesh Size -20 Mesh
Chemical Name or Material Iron powder
Product Identifier
  • Iron powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'