missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron phosphide, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Iron phosphide is used as a semiconductor. It finds application in high power and high frequency applications like laser diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Gray |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.5% (Metals basis) |
Solubility Information | Soluble in nitric acid,hydrogen fluoride and aqua regia. Insoluble in water,dilute acid and alkalis. |
Formula Weight | 198.51 |
Density | 6.74g/cm3 at 20°C |
Chemical Name or Material | Iron phosphide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'