missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(0) pentacarbonyl, Thermo Scientific Chemicals
21.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 103.0°C |
Melting Point | -20.0°C |
Color | Brown to Orange-Yellow |
Physical Form | Liquid |
Quantity | 25 g |
Assay Percent Range | 99.50% |
Linear Formula | Fe(CO)5 |
Fieser | 01,519; 02,229; 03,167; 04,268; 05,357; 06,304; 10,221; 13,152; 17,270 |
Flash Point | −15°C |
Merck Index | 15, 5143 |
Show More |
Safety and Handling
Product Identifier
- Iron pentacarbonyl
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 1
- H330-Fatal if inhaled.
- H372-Causes damage to organs.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'