missing translation for 'onlineSavingsMsg'
Learn More

Iron Nickel powder, greater-than99.5% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 036683.14

Code : NEW

Additional Details : Weight : 0.02500kg Transport : UN number : 3089 Chem class : 4.1 Pack group : II


Quantity 25g

 View more versions of this product

Product Code. 15466165

  • £26.70 / 25g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket


Specifications

Chemical Identifiers

Iron Nickel powder
>99.5% (metals basis)
MFCD01741928
Powder
Ni:Fe; 50:50 wt%

Specifications

Iron Nickel powder
Silver gray powder
>99.5% (metals basis)
MFCD01741928
Silver-gray
25g
Powder
Ni:Fe; 50:50 wt%
Odorless
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iron Nickel powder

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1

Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains¬†: Nickel, Iron

SDS