Learn More
Thermo Scientific Alfa Aesar Iron naphthenate, typically 38% in mineral spirits (6% Fe)
Brand: Thermo Scientific Alfa Aesar 043752.A9
Iron naphthenate, typically 38% in mineral spirits (6% Fe)
Code : NEW
Additional Details : CAS Number : 1338-14-3 Weight : 10.00000kg Transport : UN number : 1993 Chem class : 3 Pack group : III