Learn More
Thermo Scientific Alfa Aesar Iron naphthenate, 80% in mineral spirits (12% Fe)
Brand: Thermo Scientific Alfa Aesar 038590.36
Iron naphthenate, 80% in mineral spirits (12% Fe)
Code : NEW
Additional Details : CAS Number : 1338-14-3 Weight : 0.50000kg Transport : UN number : 1993 Chem class : 3 Pack group : III