missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iron naphthenate, 80% in mineral spirits (12% Fe)

Brand:  Alfa Aesar™ 038590.A3

Code : NEW

Additional Details : CAS Number : 1338-14-3 Weight : 2.00000kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 2kg

 View more versions of this product

Product Code. 11398897

  • £103.67 / 2kg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

Iron naphthenate
1338-14-3
MFCD00147295
UN1993

Specifications

Iron naphthenate
Liquid
MFCD00147295
UN1993
2kg
1338-14-3
1.05g/mL
80% in min.eral spirits (12% Fe)
Brown
>38° (100°F)
Videos
SDS