missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number 11317648
Click to view available options
Quantity:
250 g
1 kg
5 kg
15244-10-7
Fe2H2O13S3
417.87
MFCD00011007
VXWSFRMTBJZULV-UHFFFAOYSA-H
167265
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
15244-10-7
Fe2H2O13S3
417.87
MFCD00011007
VXWSFRMTBJZULV-UHFFFAOYSA-H
167265
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray-White
Physical Form Granules, Powder
Quantity 250 g
Solubility Information Soluble in waterSoluble in water. Sparingly soluble in alcohol.
IUPAC Name diiron(3+) hydrate trisulfate
Formula Weight 399.87 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Appearance Grayish-white powder or granules
Density 2.0-2.1
Chemical Name or Material Iron(III) sulfate hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Iron(III) sulfate hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'