missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
25 kg
6 x 1 kg
Packaging:
Paper / Poly Bag
Poly Bottle
Unit Size:
1kg
25kg
500g
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| CAS Max % | 98.0000%, 2.0000% |
| Quantity | 6 x 1 kg |
| Linear Formula | Fe2(SO4)3 · xH2O |
| UN Number | UN3260 |
| Formula Weight | 399.88 |
| Percent Purity | 21 to 23% (Fe basis) |
| Grade | puriss. p.a. |
| Packaging | Poly Bottle |
| Chemical Name or Material | Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction