missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, magnetic, Alfa Aesar™, NanoArc™

Nanoarc Magnetic Iron Oxide Is a Highly Magnetic Material in Which the Crystallites Are >98% Gamma-pHase. This Material Is Suitable For a Variety of Applications Requiring Strong Magnetic Properties and Narrow Particle

Brand:  Alfa Aesar 044896.22

Code : NEW

Additional Details : CAS Number : 1309-37-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11470870

  • £135.20 / 100g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Iron(III) oxide, magnetic
Powder
159.69
1309-37-1
Fe2O3
MFCD00011008

Specifications

Iron(III) oxide, magnetic
Powder
Red-brown powder
MFCD00011008
Red-brown
1309-37-1
Fe2O3
159.69
Odorless
100g
Videos
SDS