missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) nitrate nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
40.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 47.0°C |
Color | Gray to Purple |
Physical Form | Adhering Crystalline Powder |
Quantity | 500 g |
Assay Percent Range | 98+% |
Linear Formula | Fe(NO3)3·9H2O |
UN Number | 1466 |
Merck Index | 15, 4054 |
Formula Weight | 404 |
Percent Purity | ≥98% |
Show More |
Safety and Handling
Product Identifier
- Iron(III) nitrate nonahydrate
- Warning
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'