missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) fluoride trihydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
The primary commercial use of iron(III) fluoride in the production of ceramics. Some cross coupling reaction are catalyzed by ferric fluoride-based compounds. Iron(III) fluoride has also been shown to catalyze chemoselective addition of cyanide to aldehydes to give the cyanohydrins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Brown |
Physical Form | Crystalline |
Quantity | 50 g |
UN Number | UN3260 |
Merck Index | 14,4022 |
Solubility Information | Slightly soluble in water. Sparingly soluble in alcohol,ether,benzene. |
Formula Weight | 166.89 (112.84 Anhydrous) |
Sensitivity | Hygroscopic |
Chemical Name or Material | Iron(III) fluoride trihydrate |
Safety and Handling
Product Identifier
- Iron(III) fluoride trihydrate
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'