missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) ethoxide, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051729
Click to view available options
Quantity:
0.5 g
2 g
10 g
50 g
This item is not returnable. View return policy
5058-42-4
C6H15FeO3
191.028
MFCD00078028
JTPUGUWXHGEEHW-UHFFFAOYSA-N
iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg
6452222
ethanolate;iron(3+)
CC[O-].CC[O-].CC[O-].[Fe+3]
This item is not returnable. View return policy
5058-42-4
C6H15FeO3
191.028
MFCD00078028
JTPUGUWXHGEEHW-UHFFFAOYSA-N
iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg
6452222
ethanolate;iron(3+)
CC[O-].CC[O-].CC[O-].[Fe+3]

Precursor for Sol-Gel processes

Precursor for Sol-Gel processes.Iron(III) ethoxide is used as a precursor for inorganic compound iron oxide. It is used in the preparation of iron-platinum nanoparticles by pyrolysis with platinum(II) acetylacetonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Melting Point 120°C
Color Brown to Gray
Boiling Point 155°C (0.1 mmHg)
Odor Odorless
Linear Formula Fe(OCH2CH3)3
Quantity 0.5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Soluble in anhydrous hexane,alcohols;Miscible with hexane,benzene and ethanol.
Formula Weight 191.03
Percent Purity 99.6%
Physical Form Chunks and/or Powder
Assay (metals basis)
Chemical Name or Material Iron(III) ethoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) ethoxide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'