missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals

Catalog Number p-3257021
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10025-77-1
Cl3FeH12O6
270.29
MFCD00149712
NQXWGWZJXJUMQB-UHFFFAOYSA-K
Ferric chloride hexahydrate
iron(3+) hexahydrate trichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10025-77-1
Cl3FeH12O6
270.29
MFCD00149712
NQXWGWZJXJUMQB-UHFFFAOYSA-K
Ferric chloride hexahydrate
iron(3+) hexahydrate trichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Used to prepare organically templated iron phosphates.Also, used for sewerage treatment of industrial wastes, chloride hydrometallurgy. Useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications:
  • Used to prepare organically templated Iron Phosphates
  • Used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin) process for production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards
  • Plays important role in photoengraving and photography as an etching agent
  • Usedl as mordant in dyeing and printing textiles.
  • Also usedl in detection of phenols and phenol derivatives; gamma-hydroxybutyric acids, in Trinder spot test for detecting salicylic acids
TRUSTED_SUSTAINABILITY
Boiling Point 280°C to 285°C
Melting Point 37°C
Color Brown-Yellow
Physical Form Lump
Quantity 25 g
Assay Percent Range 97.0 to 102.0%
UN Number UN3260
Solubility Information Freely soluble in water, alcohol, ether, acetone.Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.
Formula Weight 270.30 (162.21 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow-Brown lump
Density 1.820 g/cm3
Chemical Name or Material Iron(III) Chloride Hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'