missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) bromide, anhydrous, 98+%, Thermo Scientific Chemicals

Catalog Number p-7049913
Click to view available options
Quantity:
5 g
100 g
10031-26-2
MFCD00016082
10031-26-2
MFCD00016082

Iron(III) bromide is used as a Lewis acid catalyst in the halogenation of aromatic compounds. It is also used in the preparation of 2,5-dibromo-4-aryl-1,3-pentadiene by reacting with homoallenic bromohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color Red
Physical Form Crystalline
Quantity 5 g
Merck Index 14,4018
Solubility Information Soluble in water,ether and alcohol.
Formula Weight 295.57
Percent Purity ≥98%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Iron(III) bromide, Anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) bromide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'