missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) acrylate, 96%, Thermo Scientific™

Catalog Number p-7051800
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
55488-18-1
C9H9FeO6
269.01
MFCD00080380
WNWBIDPJHFYYLM-UHFFFAOYSA-K
iron iii acrylate, feracryl, pk 2 acrylic polymer, poly acrylic acid iron salt, iron acrylate, 2-propenoic acid, homopolymer, iron salt, iron triacrylate, 79-10-7 parent, iron 3+ acrylate, 2-propenoic acid, iron 3+ salt
171450
iron(3+);prop-2-enoate
C=CC(=O)[O-].C=CC(=O)[O-].C=CC(=O)[O-].[Fe+3]
This item is not returnable. View return policy
55488-18-1
C9H9FeO6
269.01
MFCD00080380
WNWBIDPJHFYYLM-UHFFFAOYSA-K
iron iii acrylate, feracryl, pk 2 acrylic polymer, poly acrylic acid iron salt, iron acrylate, 2-propenoic acid, homopolymer, iron salt, iron triacrylate, 79-10-7 parent, iron 3+ acrylate, 2-propenoic acid, iron 3+ salt
171450
iron(3+);prop-2-enoate
C=CC(=O)[O-].C=CC(=O)[O-].C=CC(=O)[O-].[Fe+3]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 190°C to 200°C (decomposition)
Quantity 5 g
Sensitivity Air and moisture sensitive
Formula Weight 269.01
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Iron(III) acrylate
compliance-icons
Product Identifier
  • Iron(III) acrylate
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'