missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) titanium oxide, 99.8+% (metals basis), Thermo Scientific Chemicals

Catalog Number 11312238
Click to view available options
Quantity:
50 g
250 g
12022-71-8
MFCD00064704
Iron(II) titanate
12022-71-8
MFCD00064704
Iron(II) titanate

Iron(II) titanium oxide is used as a chief source of titanium. It is also used to prepare titanium dioxide by reacting sulfuric acid to get titanium oxygen sulfate followed by hydrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 151.75
Percent Purity ≥99.8%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Iron(II) titanium oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'