missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
CAS Max % | 100.0000% |
Quantity | 25 kg |
Assay Percent Range | 99.5 to 104.5% |
Linear Formula | FeSO4 · 7H2O |
Formula Weight | 278.01 |
Percent Purity | 99.5 to 104.5% (manganometric) |
Grade | puriss. |
Packaging | Paper / Poly Bag |
Vapor Pressure | 14.6mmHg (25°C) |
Density | 1.898 g/mL at 25 °C(lit.) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'