missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfate Heptahydrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073099
Click to view available options
Quantity:
2.5 kg
5 kg
7782-63-0
FeH14O11S
278.01
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
62662
CHEBI:75836
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
7782-63-0
FeH14O11S
278.01
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
62662
CHEBI:75836
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
Melting Point 64°C
Color Blue-Green
pH 3 to 4
Physical Form Solid
Quantity 2.5 kg
IUPAC Name λ²-iron(2+) heptahydrate sulfate
Formula Weight 278.01g/mol
Packaging Plastic powder jar
Vapor Pressure 14.6mmHg at 25°C
compliance-icons
Product Identifier
  • Iron(II) sulfate heptahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iron (II) Sulfate heptahydrate, Ferroussulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'