missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™

Brand:  Fisher Chemical I/1157/60

Code : 22

Additional Details : CAS Number : 7782-63-0 Weight : 1.00000kg


Quantity 1kg

 View more versions of this product

Product Code. 10161570

  • £133.00 / 1kg

In stock, ready to ship: 62

Please to purchase this item. Need a web account? Register with us today!


For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Description

Description

Specifications

Chemical Identifiers

7782-63-0
278.006
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
iron(2+);sulfate;heptahydrate
FeH14O11S
149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]

Specifications

7782-63-0
149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron(2+);sulfate;heptahydrate
62662
278.01g/mol
Plastic powder jar
Blue-Green
3 to 4
FeH14O11S
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
278.006
CHEBI:75836
Solid
14.6mmHg at 25°C
64°C
1kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iron(II) sulfate heptahydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Iron (II) Sulfate heptahydrate, Ferroussulfate

SDS Documents
Certificates