missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
Description
It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 190°C (decomposition) |
Physical Form | Powder |
Quantity | 500 g |
Solubility Information | Soluble in water. Insoluble in acetic acid. |
IUPAC Name | λ²-iron(2+) dihydrate oxalate |
Formula Weight | 179.90 (143.87 Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Sensitivity | Hygroscopic |
Density | 2.28 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'