missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11320999
61.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6047-25-2
C2H4FeO6
179.89
MFCD00150040 MFCD00012475
NPLZZSLZTJVZSX-UHFFFAOYSA-L
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™
516788
O.O.[Fe++].[O-]C(=O)C([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6047-25-2
C2H4FeO6
179.89
MFCD00150040 MFCD00012475
NPLZZSLZTJVZSX-UHFFFAOYSA-L
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™
516788
O.O.[Fe++].[O-]C(=O)C([O-])=O

It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190°C (decomposition)
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water. Insoluble in acetic acid.
IUPAC Name λ²-iron(2+) dihydrate oxalate
Formula Weight 179.90 (143.87 Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Sensitivity Hygroscopic
Density 2.28 g/mL
Assay (metals basis)
Chemical Name or Material Iron(II) oxalate dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'