missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iron(II,III) oxide, Puratronic™, 99.997% (metals basis)

Brand:  Alfa Aesar™ 012962.22

Code : NEW

Additional Details : CAS Number : 1317-61-9 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11372048

  • £428.00 / 100g


Estimated Shipment date
from Supplier 27-04-2018
Add to basket

Specifications

Chemical Identifiers

Iron(II,III) oxide
Puratronic™
MFCD00011010
1317-61-9
231.54
14,4040

Specifications

Iron(II,III) oxide
Puratronic™
99.997%
231.54
14,4040
Odorless
Black
100g
1317-61-9
Powder
(metals basis)
MFCD00011010
5.2g/mL
3.0
1538°C
Videos
SDS