missing translation for 'onlineSavingsMsg'
Learn More

Iron(II,III) oxide, 97% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 012374.A7

Code : NEW

Additional Details : CAS Number : 1317-61-9 Weight : 5.00000kg

Quantity 5kg

 View more versions of this product

Product Code. 11311769

  • £89.40 / 5kg


EU Stock 4
Add to basket

Specifications

Chemical Identifiers

Iron(II, III) oxide
-325 Mesh Powder
Fe3O4
MFCD00011010
1317-61-9
97% (metals basis)
231.54
Ferrosoferric oxide, Ferric ferrous oxide

Specifications

Iron(II, III) oxide
-325 Mesh Powder
Fe3O4
MFCD00011010
5.2g/mL
1317-61-9
97% (metals basis)
231.54
Ferrosoferric oxide, Ferric ferrous oxide
5kg
Videos
SDS