missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride tetrahydrate, 99+%, Thermo Scientific Chemicals

Catalog Number 11994181
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
13478-10-9
Cl2Fe·4H2O
198.81
WSSMOXHYUFMBLS-UHFFFAOYSA-L
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
16211588
CHEBI:86249
dichloroiron;tetrahydrate
O.O.O.O.Cl[Fe]Cl
13478-10-9
Cl2Fe·4H2O
198.81
WSSMOXHYUFMBLS-UHFFFAOYSA-L
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
16211588
CHEBI:86249
dichloroiron;tetrahydrate
O.O.O.O.Cl[Fe]Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 105.0°C to 110.0°C
Color Yellow to Green
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 5 kg
Assay Percent Range 99+%
Linear Formula FeCl2·4H2O
Merck Index 14,4043
Solubility Information Solubility in water: 1600g/L (10°C) 4100g/L (100°C). Other solubilities: soluble in alcohol
Formula Weight 198.81
Percent Purity ≥99%
Packaging Plastic bottle
Chemical Name or Material Iron(II) chloride tetrahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) chloride tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'