missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7058923
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
iron(2+);dibromide
[Fe++].[Br-].[Br-]
This item is not returnable. View return policy
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
iron(2+);dibromide
[Fe++].[Br-].[Br-]

It is used as a Polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 684°C
Physical Form Powder
Quantity 25 g
UN Number UN3260
Merck Index 14,4042
Solubility Information It is soluble in water and other solvents such as THF,methanol,ethanol.
Formula Weight 215.67
Percent Purity 99.995%
Odor Odorless
Packaging Ampule
Sensitivity Air sensitive; Hygroscopic
Density 4.636 g/mL
Assay (metals basis)
Chemical Name or Material Iron (II) bromide, Ultra dry
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) bromide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'