missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, anhydrous, 98% min, Thermo Scientific Chemicals

Catalog Number p-4407470
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
λ²-iron(2+) dibromide
[Fe++].[Br-].[Br-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-46-0
Br2Fe
215.65
MFCD00016081
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
82240
λ²-iron(2+) dibromide
[Fe++].[Br-].[Br-]

Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 684°C
Color Yellow
Physical Form Granules
Quantity 5 g
Assay Percent Range 98% min.
Solubility Information Soluble in water,methanol,ethanol and tetrahydrofuran.
Formula Weight 215.67
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Yellow granules
Density 4.636 g/cm3
Chemical Name or Material Iron(II) bromide, anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) bromide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'