missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iron foil, 0.25mm (0.01 in.) thick, Puratronic™, 99.995% (metals basis)

Brand:  Alfa Aesar™ 011380.FI

Code : NEW

Additional Details : CAS Number : 7439-89-6 Weight : 0.01000kg

Quantity 50x50mm

 View more versions of this product

Product Code. 11305849

  • £153.10 / Each


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Iron foil
0.25mm (0.01 in.) thick
MFCD00010999
7439-89-6
Puratronic™

Specifications

Iron foil
50x50mm
7439-89-6
Foil
(metals basis)
≈1.23g/25x25mm
0.25mm (0.01 in.) thick
Puratronic™
99.995%
MFCD00010999
Videos
SDS