Learn More
Iridium standard solution, for AAS, 1 mg/ml Ir in 5% HCl, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196045000
Code : 29
Additional Details : CAS Number : 7647-01-0 Weight : 0.51000kg Transport : UN number : 1789 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Iridium standard solution | |
13569-57-8,7647-01-0,7732-18-5 | |
Pink | |
LNJXVUXPFZKMNF-UHFFFAOYSA-K | |
trichloroiridium;trihydrate | |
6102153 | |
1.0200g/mL |
990 to 1010μg/mL of Ir(3+) in 10 to 20% HCl | |
500 mL | |
Cl3H6IrO3 | |
O.O.O.Cl[Ir](Cl)Cl | |
352.612 | |
AAS | |
Solution |
Safety and Handling
- Iridium atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only