missing translation for 'onlineSavingsMsg'
Learn More

Iridium plasma standard solution, Ir 10,000μg/mL, Alfa Aesar™, Specpure™

Brand:  Alfa Aesar 035752.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 1789 Chem class : 1a Pack group : II

Quantity 100mL

 View more versions of this product

Product Code. 11308637

  • £295.00 / 100mL


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Iridium
Liquid
IrCl3 in 20% HCl
MFCD00011062

Specifications

Iridium
Liquid
MFCD00011062
100mL
Liquid
IrCl3 in 20% HCl
Characteristic
Videos
SDS