missing translation for 'onlineSavingsMsg'
Learn More

Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4408555
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
7439-88-5
Ir
192.22
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
CHEBI:49666
iridium
[Ir]
This item is not returnable. View return policy
7439-88-5
Ir
192.22
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
CHEBI:49666
iridium
[Ir]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Odor Characteristic
Physical Form Liquid
Chemical Name or Material Iridium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'