missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iridium, 5% on calcium carbonate powder

Selective hydrogenation of olefins to alkanes, carbonyls to alcohols

Brand:  Alfa Aesar™ 041305.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11359677

  • £28.41 / 5g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

Iridium
5% on calcium carbonate powder
MFCD00011062

Specifications

Iridium
Powder
Odorless
5% on calcium carbonate powder
MFCD00011062
5g
Videos
SDS