missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iridium, 5% on calcium carbonate powder

Selective hydrogenation of olefins to alkanes, carbonyls to alcohols

Brand:  Alfa Aesar™ 041305.22

Code : NEW

Additional Details : Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11339677

  • £466.19 / 100g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

Iridium
5% on calcium carbonate powder
MFCD00011062

Specifications

Iridium
Powder
Odorless
5% on calcium carbonate powder
MFCD00011062
100g
Videos
SDS