missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iridium, 1% on 5mm alumina spheres, reduced, anhydrous

Brand:  Alfa Aesar™ 045570.09

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15477005

  • £85.60 / 10g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Iridium
1% on 5mm alumina spheres, Reduced, Anhydrous
MFCD00011062

Specifications

Iridium
1% on 5mm alumina spheres, Reduced, Anhydrous
MFCD00011062
10g
Videos
SDS