missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Iridium, 1% on 5mm alumina spheres, reduced, anhydrous

Brand:  Alfa Aesar™ 045570.18

Code : NEW

Additional Details : Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 15487005

  • £358.40 / 50g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Iridium
1% on 5mm alumina spheres, Reduced, Anhydrous
MFCD00011062

Specifications

Iridium
1% on 5mm alumina spheres, Reduced, Anhydrous
MFCD00011062
50g
Videos
SDS