Learn More
IRF8 Recombinant Rabbit Monoclonal Antibody (24H13L20), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 702323
Additional Details : Weight : 0.01000kg
Beschreibung
This antibody is predicted to react with Monkey, Dog and Rabbit Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
IRF8 Recombinant Monoclonal antibody specifically detects IRF8 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
IRF8 | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q02556 | |
IRF8 | |
Peptides corresponding to Human IRF8 (aa 261-277, 411-426) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
24H13L20 | |
Unconjugated | |
IRF8 | |
AI893568; H-ICSBP; Icsbp; Icsbp1; IMD32A; IMD32B; interferon consensus sequence binding protein 1; Interferon consensus sequence-binding protein; Interferon regulatory factor 8; Irf8; IRF-8; Myls; transcription factor ICSBP | |
Rabbit | |
Protein A | |
RUO | |
3394 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only