Learn More
IRF4 Recombinant Polyclonal Antibody (16 HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 711295
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Cat and Bat Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
IRF4 Recombinant Polyclonal antibody specifically detects IRF4 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
IRF4 | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q15306 | |
IRF4 | |
Peptides corresponding to Human IRF4 (aa 425-442, 175-192) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
16 HCLC | |
Unconjugated | |
IRF4 | |
AI385587; interferon regulatory factor 4; Irf4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; multiple myeloma oncogene 1; MUM1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; SHEP8; Spip; Transcriptional activator PIP | |
Rabbit | |
Protein A | |
RUO | |
3662 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |