missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IRDye™ 800CW, DBCO
Click to view available options
:
500 μg
Description
Key information: IRDye™ 800CW, DBCO is a near-infrared (NIR) fluorescent dye, supplied with a DBCO reactive group for conjugation. DBCO reactive group can react with azide-tagged molecules or biomolecules via copper-free click chemistry.
Application: Fluorescence microscopy.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 778 nm and 794 nm, respectively.
It is recommended to dissolve this dye in water or DMSO (10 - 20 mg/mL)
Specifications
Specifications
| CAS | 1373928-39-2 |
| Quantity | 500 μg |
| Target | Fluorescent Dyes |
| Molecular Formula | C64H65N4Na3O15S4 |
| Purity | 0.8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction