Learn More
Biochrom IQ/OQ Documentation for the Ultrospec 7000/8000/9000 UV-Vis Spectrophotometers
A set of Installation Qualification/Operational Qualification documents following approved US and European Pharmacopeia guidelines for wavelength accuracy, wavelength repeatability, photometric accuracy, stray light, resolution, noice and drift. Can only be performed by a trained service engineer.
Brand: Biochrom 1115701688
Code : NEW
Additional Details : Weight : 0.00001kg
Description
- Validation for our 21 CFR part 11 compliant PC software
- Robust logbook to store your instrument histroy through its life
- Flexible test templates referencing certified calibration standards
Specifications
Biochrom Ultrospec 7000/8000/9000 UV-Vis Spectrophotometers | |
IQ/OQ Documentation for the Ultrospec 7000/8000/9000 UV-Vis Spectrophotometers | |
Service |
For research use only