missing translation for 'onlineSavingsMsg'
Learn More

Ipsapirone, Tocris Bioscience™

Selective 5-HT1A agonist

Brand:  Tocris (Bio-Techne) 186950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13440154

  • £349.81 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective 5-HT1A receptor agonist (Ki = 10 nM). Anxiolytic in vivo.
Specifications

Chemical Identifiers

95847-70-4
401.485
ipsapirone, ipsapironum latin, ipsapirona spanish, ipsapirone inn:ban, unii-6j9b11mn0k, chembl8412, 2-4-4-2-pyrimidinyl-1-piperazinyl butyl-1,2-benzisothiazol-3 2h-one 1,1-dioxide, dsstox_cid_25688, dsstox_rid_81060, dsstox_gsid_45688
1,1-dioxo-2-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-1,2-benzothiazol-3-one
C19H23N5O3S
TZJUVVIWVWFLCD-UHFFFAOYSA-N
56971
C1CN(CCN1CCCCN2C(=O)C3=CC=CC=C3S2(=O)=O)C4=NC=CC=N4

Specifications

Ipsapirone
95847-70-4
ipsapirone, ipsapironum latin, ipsapirona spanish, ipsapirone inn:ban, unii-6j9b11mn0k, chembl8412, 2-4-4-2-pyrimidinyl-1-piperazinyl butyl-1,2-benzisothiazol-3 2h-one 1,1-dioxide, dsstox_cid_25688, dsstox_rid_81060, dsstox_gsid_45688
C1CN(CCN1CCCCN2C(=O)C3=CC=CC=C3S2(=O)=O)C4=NC=CC=N4
401.485
401.48
50mg
C19H23N5O3S
TZJUVVIWVWFLCD-UHFFFAOYSA-N
1,1-dioxo-2-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-1,2-benzothiazol-3-one
56971
>99%
Videos