Learn More
Tocris Bioscience™ IPA 3
Group I PAK inhibitor
Brand:  Tocris Bioscience™ 362210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463304
  • £118.80 / 10mg
Estimated Shipment
04-04-2019
Add to basket
Description
Description
Group I p21-activated kinase (PAK) inhibitor (IC 50 = 2.5 μM at PAK1). Targets the autoregulatory mechanism and promotes the inactive conformation of PAKs. Inhibits PAK1-mediated signaling in vivo; potential anti-tumor agent. Negative Control also available.
Specifications
Specifications
IPA 3
>96
10mg
IPA 3
350.45