IOX 2, Tocris™

Potent, selective HIF-1α prolyl hydroxylase-2 (PHD2) inhibitor

Overview
Brand: Thermo Scientific Nunc

Manufacturer Part Number: 445110

UNSPSC: 41113036

Code: 76

Additional Details:
Additional Details: Weight: 3.24400kg


Quantity 10mg

Product Code. 10550962

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Chemical Identifiers

Chemical Name or Material IOX 2
Assay Percent Range >99%
Molecular Formula C19H16N2O5
Formula Weight 352.34
Synonym N-[[1, 2-Dihydro-4-hydroxy-2-oxo-1-(phenylmethyl)-3-quinolinyl]carbonyl]glycine

Specification

Formula Weight 352.34
Quantity 10mg
Recommended Storage Store at +4°C
CAS 931398-72-0
Chemical Name or Material IOX 2
Assay Percent Range >99%
Synonym N-[[1, 2-Dihydro-4-hydroxy-2-oxo-1-(phenylmethyl)-3-quinolinyl]carbonyl]glycine
Molecular Formula C19H16N2O5
Documentation

Technical documents

Safety and Handling

Store at +4°C