missing translation for 'onlineSavingsMsg'
Learn More

β-Ionone, 96%, synthetic, Thermo Scientific Chemicals

Catalog Number p-148963
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14901-07-6
C13H20O
192.30
MFCD00001549
PSQYTAPXSHCGMF-BQYQJAHWSA-N
beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
638014
CHEBI:32325
(E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
CC(=O)\C=C\C1=C(C)CCCC1(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14901-07-6
C13H20O
192.30
MFCD00001549
PSQYTAPXSHCGMF-BQYQJAHWSA-N
beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
638014
CHEBI:32325
(E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
CC(=O)\C=C\C1=C(C)CCCC1(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Density 0.9440g/mL
Boiling Point 237.0°C to 239.0°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5190 to 1.5215
Quantity 5 g
Beilstein 07, 167
Merck Index 10, 4914
Specific Gravity 0.944
Solubility Information Solubility in water: slightly soluble. Other solubilities: solutle in 2 to 3 parts of 70% alcohol, ether,, chloroform and benzene, miscible with absolute alcohol
Formula Weight 192.3
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material β-Ionone, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'