Learn More
Fisherbrand™ Ion Selective Electrode
Combined ion-sensitive electrode with solid membrane interchangeable flat surface where ideally calibration should be carried out as often as possible (Less than once a day).
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- Combined ion-sensitive electrode with solid membrane interchangeable flat surface
- The ion to be detected passes through the membrane generating a difference of Potential proportional to ionic concentration
- Analysis time typically less than one minute
- Sample preparation steps such as filtering or distillation rarely necessary
- Calibration should be carried out as often as possible (Less than once a day)
Specifications
Ion Selective Electrode |