Phosphate-P Standard for Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Overview
Brand: SPEX Certiprep

Manufacturer Part Number: 601

100 ML PHOSPHATE STANDARD SOLUTION (POTASSIUMDihydrogen Orthophosphate, KH2PO4) SPEX CertiPrep

UNSPSC: 41116107

Code: 04

Additional Details:
Additional Details: Weight: 0.14000kg


Packaging Triple-leached LDPE bottle
Quantity 125mL

Disclaimers: Product intended for laboratory use only
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Product Code. 11450189

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification

Chemical Identifiers

Chemical Name or Material Phosphate-Phosphorus Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Specification

Quantity 125mL
Concentration or Composition (by Analyte or Components) 10002g/mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Chemical Name or Material Phosphate-Phosphorus Standard
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Nonhazardous
Solvent or Matrix H2O
CAS Min % 0.1
CAS PO4-P
CAS Min % 99.9
CAS Max % 0.1
Certificate of Analysis With Certificate of Analysis
CAS Max % 99.9
CAS 7732-18-5

AnalyteConcentration
PO&subscript_4;-P1000μg/mL

Compliance: GHS/OSHA Complaint

Certifications: Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Alternative Products
Documentation Safety and Handling

Ambient